Learn More
NEW ENGLAND BIOGROUP LUNA BF-II 2nd generation validation slide. For use with LUNA-FX7 and LUNA-BX7 automated cell counters.
SDP
Catalog No. NC3903972
Encompass
LUNA BF-II 2nd generation validation slide. For use with LUNA-FX7 and LUNA-BX7 automated cell counters. The BF-II validation slide is a patterned slide and in semi-permanent use. It allows you to validate brightfield cell counting performance of the following instruments and can be used for daily quality control purpose. Does not expire.
Catalog No. NC3903972 Supplier NEW ENGLAND BIOGROUP Supplier No. L72041
LUNA BF-II 2nd generation validation slide. For use with LUNA-FX7 and LUNA-BX7 automated cell counters. The BF-II validation slide is a patterned slide and in semi-permanent use. It allows you to validate brightfield cell counting performance of the following instruments and can be used for daily quality control purpose. Does not expire.