Learn More
LOGOS BIOSYSTEMS INC LUNA™ Cell Counting Slides, 500 Slides
SDP
Catalog No. NC1762048
Encompass
The disposable LUNA™ Cell Counting Slides offer the ultimate counting experience. With no mess or cleanup, these precision slides are convenient and maintain the highest standard of cell counting accuracy. LUNA™ Cell Counting Slides are for brightfield cell counting with the automated brightfield cell counters of the LUNA™ family.
Catalog No. NC1762048 Supplier LOGOS BIOSYSTEMS INC Supplier No. L12002
The disposable LUNA™ Cell Counting Slides offer the ultimate counting experience. With no mess or cleanup, these precision slides are convenient and maintain the highest standard of cell counting accuracy. LUNA™ Cell Counting Slides are for brightfield cell counting with the automated brightfield cell counters of the LUNA™ family.