Learn More
NEW ENGLAND BIOGROUP LUNA Erythrosin B Non Toxic Cell Viability Stain, 2 x 1mL. Good for 200 tests/cell counts. For LUNA-II, LUNA-III, LUNA-FL, LUNA-BX7 and LUNA-FX7.
SDP
Catalog No. NC3903962
Encompass
LUNA Erythrosin B Non Toxic Cell Viability Stain, 2 x 1mL. Good for 200 tests/cell counts. For LUNA-II, LUNA-III, LUNA-FL, LUNA-BX7 and LUNA-FX7.
Catalog No. NC3903962 Supplier NEW ENGLAND BIOGROUP Supplier No. L13002
LUNA Erythrosin B Non Toxic Cell Viability Stain, 2 x 1mL. Good for 200 tests/cell counts. For LUNA-II, LUNA-III, LUNA-FL, LUNA-BX7 and LUNA-FX7.