Learn More
NEW ENGLAND BIOGROUP LUNA Erythrosin B Non Toxic Cell Viability Stain, 50 x 1mL. Good for 5000 tests/cell counts. For LUNA-II, LUNA-III, LUNA-FL, LUNA-BX7 and LUNA-FX7.
SDP
Catalog No. NC3903963
Encompass
LUNA Erythrosin B Non Toxic Cell Viability Stain, 50 x 1mL. Good for 5000 tests/cell counts. For LUNA-II, LUNA-III, LUNA-FL, LUNA-BX7 and LUNA-FX7.
Catalog No. NC3903963 Supplier NEW ENGLAND BIOGROUP Supplier No. L13003
LUNA Erythrosin B Non Toxic Cell Viability Stain, 50 x 1mL. Good for 5000 tests/cell counts. For LUNA-II, LUNA-III, LUNA-FL, LUNA-BX7 and LUNA-FX7.