Learn More
NEW ENGLAND BIOGROUP LUNA Fluorescent validation slide. For use with LUNA-FX7 automated cell counters for quality control in fluorescent mode. One year shelf life.
SDP
Catalog No. NC3903974
Encompass
LUNA Fluorescent validation slide. For use with LUNA-FX7 automated cell counters for quality control in fluorescent mode. One year shelf life. The LUNA FL Validation Slide is a slide with pre-fixed fluorescence beads and in a limited expiration period. It allows you to validate fluorescence cell counting performance of the following instruments and can be used for daily quality control purpose.
Catalog No. NC3903974 Supplier NEW ENGLAND BIOGROUP Supplier No. L72030
LUNA Fluorescent validation slide. For use with LUNA-FX7 automated cell counters for quality control in fluorescent mode. One year shelf life. The LUNA FL Validation Slide is a slide with pre-fixed fluorescence beads and in a limited expiration period. It allows you to validate fluorescence cell counting performance of the following instruments and can be used for daily quality control purpose.