Learn More
Adam Equipment Adam Equipment Luna Precision Balance LBL 34001E 34000g capacity x 0.1g readability
Catalog No. NC2612389
Encompass_Preferred
Adam Equipment Luna Precision Balances are urable and reliable, Luna large pan precision balances deliver a fine readability of 0.1g. Innovative design enables a compact footprint, allowing Luna to occupy minimal bench space while offering a high level of performance. Single-piece construction offers greater stability, producing highly repeatable results. The mountable weighing indicator combines durability and performance in a stylish design, with a vivid, crystal-clear LCD display that delivers results in bright 24mm digits against a black background. Efficiency in the mechanics added to impressive processing power of the electronics leads to excellent performance. Durable and reliable, Luna large pan precision balances deliver a fine readability of 0.1g. Innovative design with a mountable indicator enables a compact footprint, so it occupies minimal space while retaining a high level of performance. AC adapter included.
Catalog No. NC2612389 Supplier Adam Equipment Supplier No. LBL 34001E
Adam Equipment Luna Precision Balances are urable and reliable, Luna large pan precision balances deliver a fine readability of 0.1g. Innovative design enables a compact footprint, allowing Luna to occupy minimal bench space while offering a high level of performance. Single-piece construction offers greater stability, producing highly repeatable results. The mountable weighing indicator combines durability and performance in a stylish design, with a vivid, crystal-clear LCD display that delivers results in bright 24mm digits against a black background. Efficiency in the mechanics added to impressive processing power of the electronics leads to excellent performance. Durable and reliable, Luna large pan precision balances deliver a fine readability of 0.1g. Innovative design with a mountable indicator enables a compact footprint, so it occupies minimal space while retaining a high level of performance. AC adapter included.