Learn More
Adam Equipment Adam Equipment Luna Precision Balance LBL 24001E 24000g capacity x 0.1g readability
Supplier:  Adam Equipment LBL 24001E
Encompass_Preferred
Adam Equipment Luna Precision Balances are urable and reliable, Luna large pan precision balances deliver a fine readability of 0.1g. Innovative design enables a compact footprint, allowing Luna to occupy minimal bench space while offering a high level of performance. Single-piece construction offers greater stability, producing highly repeatable results. The mountable weighing indicator combines durability and performance in a stylish design, with a vivid, crystal-clear LCD display that delivers results in bright 24mm digits against a black background. Efficiency in the mechanics added to impressive processing power of the electronics leads to excellent performance. Durable and reliable, Luna large pan precision balances deliver a fine readability of 0.1g. Innovative design with a mountable indicator enables a compact footprint, so it occupies minimal space while retaining a high level of performance. AC adapter included.
Catalog No. NC2612391