Learn More
LOGOS BIOSYSTEMS INC LUNA Reusable Slide, One Chamber with One Coverslip
SDP
Catalog No. NC1842173
Encompass
The LUNA Reusable Slide may be used for both fluorescence and brightfield cell counting with the LUNA family of automated cell counters and the CELENA S Digital Imaging System.
Catalog No. NC1842173 Supplier LOGOS BIOSYSTEMS INC Supplier No. L12011
The LUNA Reusable Slide may be used for both fluorescence and brightfield cell counting with the LUNA family of automated cell counters and the CELENA S Digital Imaging System.