Learn More
LOGOS BIOSYSTEMS INC LUNA™ Standard Beads, 2 x 1 mL
SDP
Catalog No. NC1549817
Encompass
UNA™ Standard Beads are used to calibrate the LUNA™ family of cell counters for brightfield cell counting.
Catalog No. NC1549817 Supplier LOGOS BIOSYSTEMS INC Supplier No. B13101
UNA™ Standard Beads are used to calibrate the LUNA™ family of cell counters for brightfield cell counting.