Learn More
New England Biolabs, Inc. Luna Universal Probe qPCR Master Mix – 200 rxn (2 x 1 ml)
SDP
Catalog No. 501526408
Encompass
The NEB Luna Universal Probe qPCR Master Mix is a 2X reaction mix optimized for real-time qPCR detection and quantitation of target DNA sequences using hydrolysis probes. It contains Hot Start Taq DNA Polymerase and has been formulated with a unique passive reference dye that is compatible across a variety of instrument platforms (including those that require a high or low ROX reference signal). It also features dUTP for carryover prevention and a non-fluorescent, visible dye to monitor reaction setup. This dye does not spectrally overlap fluorophores commonly used for qPCR and will not interfere with real-time detection. The master mix formulation is supplied at 2X concentration and contains all PCR components required for amplification and quantitation of DNA except primers/probes and DNA template. Genomic DNA or cDNA of interest can be quantitated with Luna qPCR and existing as well as commercial qPCR assay primer/probe sequences can be used.
Catalog No. 50-152-6408 Supplier New England Biolabs, Inc. Supplier No. M3004S
The NEB Luna Universal Probe qPCR Master Mix is a 2X reaction mix optimized for real-time qPCR detection and quantitation of target DNA sequences using hydrolysis probes. It contains Hot Start Taq DNA Polymerase and has been formulated with a unique passive reference dye that is compatible across a variety of instrument platforms (including those that require a high or low ROX reference signal). It also features dUTP for carryover prevention and a non-fluorescent, visible dye to monitor reaction setup. This dye does not spectrally overlap fluorophores commonly used for qPCR and will not interfere with real-time detection. The master mix formulation is supplied at 2X concentration and contains all PCR components required for amplification and quantitation of DNA except primers/probes and DNA template. Genomic DNA or cDNA of interest can be quantitated with Luna qPCR and existing as well as commercial qPCR assay primer/probe sequences can be used.