Learn More
New England Biolabs, Inc. Luna Universal qPCR Master Mix – 1000 rxn (10 x 1 ml)
SDP
Supplier:  New England Biolabs, Inc. M3003X
Encompass
The NEB Luna Universal qPCR Master Mix is an optimized 2X reaction mix for real-time qPCR detection and quantitation of target DNA sequences using the SYBR/FAM channel of most real-time qPCR instruments. It contains Hot Start Taq DNA Polymerase and has been formulated with a unique passive reference dye that is compatible across a variety of instrument platforms (including those that require a high or low ROX reference signal). It also features dUTP for carryover prevention and a non-fluorescent, visible dye to monitor reaction setup. This dye does not spectrally overlap with fluorescent dyes used for qPCR and will not interfere with real-time detection. The master mix formulation is supplied at 2X concentration and contains all PCR components required for amplification and quantitation of DNA except primers and DNA template. Genomic DNA or cDNA of interest can be quantitated with Luna qPCR, and existing as well as commercial qPCR assay primer sequences can be used.
Catalog No. 50-152-6405