Learn More
New England Biolabs, Inc. Luna® Universal Probe One-Step RT-qPCR Kit – 500 reactions
SDP
Catalog No. NC1241930
Encompass
In the Luna Universal One-Step Probe RT-qPCR Kit, Hot Start Taq DNA Polymerase is combined with a novel WarmStart-activated reverse transcriptase, allowing dual control of enzyme activity via reversible, aptamer-based inhibition. This temperature-dependent activation helps to prevent undesirable non-specific priming and extension prior to thermocycling, providing added security for setting up reactions at room temperature. The engineered WarmStart Luna Reverse Transcriptase also possesses higher thermostability than many other RTs, allowing an optimal reaction temperature of 55°C. For difficult targets/templates, higher RT step temperatures of up to 60°C can be used without compromising Luna performance. Note that to ensure full activation of the WarmStart Luna RT, incubation at temperatures lower than 50°C is not recommended.
  • Non-interfering, visible tracking dye helps to eliminate pipetting errors
Catalog No. NC1241930 Supplier New England Biolabs, Inc. Supplier No. E3006L
In the Luna Universal One-Step Probe RT-qPCR Kit, Hot Start Taq DNA Polymerase is combined with a novel WarmStart-activated reverse transcriptase, allowing dual control of enzyme activity via reversible, aptamer-based inhibition. This temperature-dependent activation helps to prevent undesirable non-specific priming and extension prior to thermocycling, providing added security for setting up reactions at room temperature. The engineered WarmStart Luna Reverse Transcriptase also possesses higher thermostability than many other RTs, allowing an optimal reaction temperature of 55°C. For difficult targets/templates, higher RT step temperatures of up to 60°C can be used without compromising Luna performance. Note that to ensure full activation of the WarmStart Luna RT, incubation at temperatures lower than 50°C is not recommended.
  • Non-interfering, visible tracking dye helps to eliminate pipetting errors