Learn More
Phenomenex Inc Phenomenex LC Column, C18(3), 10 μm, 150 x 4.6 mm
Supplier:  Phenomenex Inc 00F4616E0
Encompass
A preparative reversed-phase HPLC column optimized for large-scale separation of hydrophobic compounds. The column uses 10 μm silica particles with a C18(3) bonded phase and a 100 Å pore size, and is sized 150 x 4.6 mm for prep LC applications.
  • Prep-scale reversed-phase C18(3) chemistry for hydrophobic analytes.
  • 10 μm particle size for high loading capacity and preparative separations.
  • 100 Å pore size suitable for small to mid-sized molecules.
  • 150 x 4.6 mm dimensions compatible with preparative HPLC systems.
  • Provides robust retention and reproducible separations for large-scale work.
Catalog No. NC3596458