Learn More
New England Biolabs, Inc. Luna Probe One-Step RT-qPCR Kit (No ROX) – 2500 reactions
SDP
Supplier:  New England Biolabs, Inc. E3007E
Encompass
In the Luna Probe One-Step RT-qPCR Kit (No ROX), Hot Start Taq DNA Polymerase is combined with a novel WarmStart-activated reverse transcriptase, allowing dual control of enzyme activity via reversible, aptamer-based inhibition. This temperature-dependent activation helps to prevent undesirable non-specific priming and extension prior to thermocycling, providing added security for setting up reactions at room temperature. The engineered WarmStart Luna Reverse Transcriptase also possesses higher thermostability than many other RTs, allowing an optimal reaction temperature of 55°C. For difficult targets/templates, higher RT step temperatures of up to 60°C can be used without compromising Luna performance. Note that to ensure full activation of the WarmStart Luna RT, incubation at temperatures lower than 50°C is not recommended. This formulation contains no reference dye and is compatible with any instrument that does not require ROX (if ROX normalization is needed, ROX can be added.)
Catalog No. 50-202-4003