Learn More
New England Biolabs, Inc. LunaScript RT Master Mix Kit (Primer-free) - 25 rxn
SDP
Catalog No. 502108544
Encompass
LunaScript RT Master Mix Kit (Primer-free) is an optimized master mix containing all the necessary components for first strand cDNA synthesis except for primers. The mix is compatible with random primers, oligo dT primers, and gene-specific primers enabling maximum cDNA synthesis flexibility. LunaScript RT Master Mix (Primer-free) features the thermostable Luna Reverse Transcriptase which supports cDNA synthesis at elevated temperatures. Murine RNase Inhibitor is included to protect template RNA from degradation. In addition, the presence of a blue tracking dye provides a visual indicator for the RT step as well as downstream applications.
Catalog No. 50-210-8544 Supplier New England Biolabs, Inc. Supplier No. E3025S
LunaScript RT Master Mix Kit (Primer-free) is an optimized master mix containing all the necessary components for first strand cDNA synthesis except for primers. The mix is compatible with random primers, oligo dT primers, and gene-specific primers enabling maximum cDNA synthesis flexibility. LunaScript RT Master Mix (Primer-free) features the thermostable Luna Reverse Transcriptase which supports cDNA synthesis at elevated temperatures. Murine RNase Inhibitor is included to protect template RNA from degradation. In addition, the presence of a blue tracking dye provides a visual indicator for the RT step as well as downstream applications.