Learn More
New England Biolabs, Inc. LunaScript RT SuperMix - 100 rxns
SDP
Catalog No. 502253076
Encompass
LunaScript RT SuperMix is an optimized master mix containing all the necessary components for first strand cDNA synthesis in the context of a two-step RT-qPCR workflow. It features the thermostable Luna Reverse Transcriptase which supports cDNA synthesis at elevated temperatures. Murine RNase Inhibitor is included to protect template RNA from degradation. LunaScript RT SuperMix contains random hexamer and poly-dT primers allowing even coverage across the length of the RNA targets. In addition LunaScript RT SuperMix contains dNTPs and a blue tracking dye providing a visual indicator that can be followed throughout the two-step RT-qPCR process. This product offers robust linear and sensitive detection using total RNA inputs as high as 1 g and as low as single copies of RNA.
This item is not returnable. View return policy
Catalog No. 50-225-3076 Supplier New England Biolabs, Inc. Supplier No. M3010L
LunaScript RT SuperMix is an optimized master mix containing all the necessary components for first strand cDNA synthesis in the context of a two-step RT-qPCR workflow. It features the thermostable Luna Reverse Transcriptase which supports cDNA synthesis at elevated temperatures. Murine RNase Inhibitor is included to protect template RNA from degradation. LunaScript RT SuperMix contains random hexamer and poly-dT primers allowing even coverage across the length of the RNA targets. In addition LunaScript RT SuperMix contains dNTPs and a blue tracking dye providing a visual indicator that can be followed throughout the two-step RT-qPCR process. This product offers robust linear and sensitive detection using total RNA inputs as high as 1 g and as low as single copies of RNA.