Learn More
New England Biolabs, Inc. LunaScript RT SuperMix Kit – 100 reactions
SDP
Catalog No. 501527876
Encompass
LunaScript RT SuperMix Kit features an optimized master mix containing all the necessary components for first strand cDNA synthesis in the context of a two-step RT-qPCR workflow. It includes the thermostable Luna Reverse Transcriptase, which supports cDNA synthesis at elevated temperatures. Murine RNase Inhibitor is also included to protect template RNA from degradation. The LunaScript RT SuperMix contains random hexamer and poly-dT primers, allowing even coverage across the length of the RNA targets. In addition, the LunaScript RT SuperMix Kit contains a blue dye, providing a visual indicator that can be followed throughout the two-step RT-qPCR process. The LunaScript RT SuperMix offers robust, linear, and sensitive detection using total RNA inputs as high as 1 µg and as low as single copies of RNA.
Catalog No. 50-152-7876 Supplier New England Biolabs, Inc. Supplier No. E3010L
LunaScript RT SuperMix Kit features an optimized master mix containing all the necessary components for first strand cDNA synthesis in the context of a two-step RT-qPCR workflow. It includes the thermostable Luna Reverse Transcriptase, which supports cDNA synthesis at elevated temperatures. Murine RNase Inhibitor is also included to protect template RNA from degradation. The LunaScript RT SuperMix contains random hexamer and poly-dT primers, allowing even coverage across the length of the RNA targets. In addition, the LunaScript RT SuperMix Kit contains a blue dye, providing a visual indicator that can be followed throughout the two-step RT-qPCR process. The LunaScript RT SuperMix offers robust, linear, and sensitive detection using total RNA inputs as high as 1 µg and as low as single copies of RNA.