Learn More
Unchained Labs LUNATIC PL FR STRIP OD40 480PK
SDP
Catalog No. NC2192266
Encompass
Lunatic Plate Frame Strips, OD 40 (Box of 480); Box of 480 Lunatic Plate Frame Strips, 0.03-40 OD 16 samplesper chip. For use only with a reusable Lunatic Plate Frame and aLunatic, DropSense96 or DropSense96 Classic platform. (Previously product code 4-238016-S)
This item is not returnable. View return policy
Catalog No. NC2192266 Supplier Unchained Labs Supplier No. 7012006
Lunatic Plate Frame Strips, OD 40 (Box of 480); Box of 480 Lunatic Plate Frame Strips, 0.03-40 OD 16 samplesper chip. For use only with a reusable Lunatic Plate Frame and aLunatic, DropSense96 or DropSense96 Classic platform. (Previously product code 4-238016-S)