Learn More
Unchained Labs Lunatic, Unchained Labs
SDP
Catalog No. NC1481311
Encompass
The Lunatic is a benchtop automated protein purification system by Unchained Labs that uses magnetic beads for affinity purification of proteins. It has a compact footprint and intuitive software interface. Key features include automated buffer mixing, customizable protocols, UV monitoring, and fraction collection. The system is designed for high-throughput purification of antibodies, enzymes, and other recombinant proteins. With its automated workflow, the Lunatic aims to simplify and accelerate protein purification.
Catalog No. NC1481311 Supplier Unchained Labs Supplier No. 7012005
The Lunatic is a benchtop automated protein purification system by Unchained Labs that uses magnetic beads for affinity purification of proteins. It has a compact footprint and intuitive software interface. Key features include automated buffer mixing, customizable protocols, UV monitoring, and fraction collection. The system is designed for high-throughput purification of antibodies, enzymes, and other recombinant proteins. With its automated workflow, the Lunatic aims to simplify and accelerate protein purification.