Learn More
Unchained Labs High Lunatic Plates, OD 200
SDP
Catalog No. NC1415573
Encompass
Box of 40 High Lunatic Plates, 0.03-200 OD. 96 samples per plate. For use with Big Lunatic, DropSense96 or DropSense96 Classic. (Previously product code 4-240096-D+)
Catalog No. NC1415573 Supplier Unchained Labs Supplier No. 7012001
Box of 40 High Lunatic Plates, 0.03-200 OD. 96 samples per plate. For use with Big Lunatic, DropSense96 or DropSense96 Classic. (Previously product code 4-240096-D+)