Learn More
Supertek™ Lung Apparatus
SCI_ED
Catalog No. S139415 Shop All Supertek Scientific Products
Change view
Click to view available options
Grade:
3+
Catalog No. Grade
S139415 3+
1 options
This item is not returnable. View return policy
Catalog No. S139415 Supplier Supertek™ Supplier No. B12116SP
Add to Cart
Add to Cart
This item is not returnable. View return policy
Hands on lung apparatus that demonstrates how the lungs are emptied and filled with air by relaxation and contraction of the diaphragm muscle.
  • Two balloons attached to a Y tube simulate the lungs.
  • The chest cavity is represented by a half-gallon plastic bell jar with an open top.
  • The stout rubber sheet acts as a diaphragm.
  • As the diaphragm is pulled downward the atmospheric pressure is lowered inside the chamber causing the lungs to expand taking in air.
  • When the diaphragm is released the lungs deflate.
  • Includes lesson plan.
Specifications
Grade 3+
Material Plastic
Product Type Lung Apparatus