Learn More
Novus Biologicals™ Lung Tissue Slides (Adenocarcinoma)- Paraffin

Description
These slides are paraffin coated to prevent sample oxidization, it is recommended that slides are first de-paraffinized by baking at 60 degrees C for 1 hour in a vertical orientation prior to performing antigen retrieval procedures.
Specifications
Specifications
| Cross Reactivity | Human |
| Type | Lung Tissue Slides (Adenocarcinoma Poorly Differentiated) |
| For Use With (Application) | Immunohistochemistry-Paraffin |
| Regulatory Status | This product is for research use only and is not approved for use in humans or in clinical diagnosis. |
| Species | Human |
| Tissue Condition | Adenocarcinoma |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.