Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Lup-20(29)-en-3-ol, 3-(hexadecanoate), (3β)- | 32214-80-5 | 25 MG
SDP

Catalog No. 5000421594
Encompass_Preferred

Lupeol palmitate is a natural compound with established antiulcer activities and a gastroprotective action.

  • Natural compound
  • Exhibits antiulcer activities
  • Provides gastroprotective action
  • Purity of 98.0%
  • Molecular weight: 665.13
  • Molecular formula: C46H80O2
  • Appearance: White to off-white solid
  • Classified as terpenoids, triterpenes, pentacyclic triterpenoids
  • Derived from plants

Catalog No. 50-004-21594 Supplier Medchemexpress LLC Supplier No. 25MGHYN3356
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Lupeol palmitate is a natural compound with established antiulcer activities and a gastroprotective action.

  • Natural compound
  • Exhibits antiulcer activities
  • Provides gastroprotective action
  • Purity of 98.0%
  • Molecular weight: 665.13
  • Molecular formula: C46H80O2
  • Appearance: White to off-white solid
  • Classified as terpenoids, triterpenes, pentacyclic triterpenoids
  • Derived from plants

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.