Learn More
Medchemexpress LLC Lup-20(29)-ene-3,28-diol, 3,28-diacetate, (3β)- | 1721-69-3 | 95.0% | 526.79 | 25 MG
SDP
Catalog No. 5000426817
Encompass_Preferred
Betulin diacetate is a triterpene, which is a derivative of Betulin. It functions as an anti-AID agent and also exhibits anti-cancer activity.
  • Triterpene
  • Derivative of betulin
  • Anti-AID agent
  • Possesses anti-cancer activity
Catalog No. 50-004-26817 Supplier Medchemexpress LLC Supplier No. 25MGHYN9437
Betulin diacetate is a triterpene, which is a derivative of Betulin. It functions as an anti-AID agent and also exhibits anti-cancer activity.
  • Triterpene
  • Derivative of betulin
  • Anti-AID agent
  • Possesses anti-cancer activity