Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Lupenone | 1617-70-5 | 5 MG
SDP

Catalog No. 5000420318
Encompass_Preferred

Lupenone is an orally active lupine-type triterpenoid isolated from *Musa basjoo*. It impacts the PI3K/Akt/mTOR and NF-κB signaling pathways, exhibiting anti-inflammatory, antiviral, antidiabetic, and anticancer properties. It protects neuroblastoma SH-SY5y cells from methamphetamine-induced apoptotic cell death and improves type 2 diabetic nephropathy in db/db mouse models.

  • Purity: 99.74%
  • Appearance: solid, white to off-white
  • Structure classification: terpenoids, triterpenes
  • Source: plants (Leguminosae *Astragalus oleifolius* DC., Musaceae *Musa basjoo* Sieb. et Zucc., Compositae)
  • Soluble in ethanol (10 mg/mL)

Catalog No. 50-004-20318 Supplier Medchemexpress LLC Supplier No. HYN25905MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Lupenone is an orally active lupine-type triterpenoid isolated from *Musa basjoo*. It impacts the PI3K/Akt/mTOR and NF-κB signaling pathways, exhibiting anti-inflammatory, antiviral, antidiabetic, and anticancer properties. It protects neuroblastoma SH-SY5y cells from methamphetamine-induced apoptotic cell death and improves type 2 diabetic nephropathy in db/db mouse models.

  • Purity: 99.74%
  • Appearance: solid, white to off-white
  • Structure classification: terpenoids, triterpenes
  • Source: plants (Leguminosae *Astragalus oleifolius* DC., Musaceae *Musa basjoo* Sieb. et Zucc., Compositae)
  • Soluble in ethanol (10 mg/mL)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.