Learn More
Selleck Chemical LLC Lupeol S3614-25mg
SDP
Catalog No. 503110525
Encompass_Preferred
Lupeol (Clerodol Monogynol B Fagarasterol Farganasterol) is a significant lupane-type triterpene represented in the plant fungi and animal kingdoms with anticancer antiprotozoal chemopreventive and anti-inflammatory properties
Catalog No. 50-311-0525 Supplier Selleck Chemical LLC Supplier No. S361425MG
Lupeol (Clerodol Monogynol B Fagarasterol Farganasterol) is a significant lupane-type triterpene represented in the plant fungi and animal kingdoms with anticancer antiprotozoal chemopreventive and anti-inflammatory properties