Learn More
Medchemexpress LLC Lupeol | 545-47-1 | 426.72 | 100 MG
SDP
Catalog No. 5000416102
Encompass_Preferred
Lupeol is an active pentacyclic triterpenoid with anti-oxidant, anti-mutagenic, anti-tumor, and anti-inflammatory activity. It is a potent androgen receptor (AR) inhibitor and can be used for cancer research, particularly for prostate cancer of androgen-dependent phenotype (ADPC) and castration-resistant phenotype (CRPC).
  • Has anti-oxidant activity
  • Has anti-mutagenic activity
  • Has anti-tumor activity
  • Has anti-inflammatory activity
  • Potent androgen receptor (AR) inhibitor
  • Can be used for cancer research
Catalog No. 50-004-16102 Supplier Medchemexpress LLC Supplier No. HYN0790100MG
Lupeol is an active pentacyclic triterpenoid with anti-oxidant, anti-mutagenic, anti-tumor, and anti-inflammatory activity. It is a potent androgen receptor (AR) inhibitor and can be used for cancer research, particularly for prostate cancer of androgen-dependent phenotype (ADPC) and castration-resistant phenotype (CRPC).
  • Has anti-oxidant activity
  • Has anti-mutagenic activity
  • Has anti-tumor activity
  • Has anti-inflammatory activity
  • Potent androgen receptor (AR) inhibitor
  • Can be used for cancer research