Learn More
Medchemexpress LLC Lupeol | 545-47-1 | 426.72 | 50 MG
SDP
Catalog No. 5000416719
Encompass_Preferred
Lupeol is an active pentacyclic triterpenoid with anti-oxidant, anti-mutagenic, anti-tumor, and anti-inflammatory activity. It is a potent androgen receptor (AR) inhibitor and is utilized for cancer research, particularly for prostate cancer of androgen-dependent phenotype (ADPC) and castration resistant phenotype (CRPC).
  • Active pentacyclic triterpenoid.
  • Anti-oxidant, anti-mutagenic, anti-tumor, and anti-inflammatory activity.
  • Potent androgen receptor (AR) inhibitor.
  • Used for cancer research, especially prostate cancer of androgen-dependent phenotype (ADPC) and castration resistant phenotype (CRPC).
Catalog No. 50-004-16719 Supplier Medchemexpress LLC Supplier No. HYN079050MG
Lupeol is an active pentacyclic triterpenoid with anti-oxidant, anti-mutagenic, anti-tumor, and anti-inflammatory activity. It is a potent androgen receptor (AR) inhibitor and is utilized for cancer research, particularly for prostate cancer of androgen-dependent phenotype (ADPC) and castration resistant phenotype (CRPC).
  • Active pentacyclic triterpenoid.
  • Anti-oxidant, anti-mutagenic, anti-tumor, and anti-inflammatory activity.
  • Potent androgen receptor (AR) inhibitor.
  • Used for cancer research, especially prostate cancer of androgen-dependent phenotype (ADPC) and castration resistant phenotype (CRPC).