Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Lupeol

Catalog No. L5632100MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mg
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L5632100MG 25 mg
L563225MG 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. L5632100MG Supplier Sigma Aldrich Supplier No. L5632100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Lupeol is a novel anti-cancer and anti-inflammatory dietary triterpene. They help in stabilizing phospholipid bilayers in plant cell membranes. In general, this group contains 28-29 carbons along with carbon-carbon double bonds, usually one in the sterol nucleus and at times a second in the alkyl side chain.Lupeol was used as reference standard in method development and validation of lupeol from Vernonia Cinerea using HPLC method. It has been used as a standard for the identification and quantification of triterpenoids in blueberry cuticular wax by GC.Tandem Mass Spectrometry data independently generated by Scripps Center for Metabolomics is available to view or download in PDF. L5632.pdf Tested metabolites are featured on Scripps Center for Metabolomics METLIN Metabolite Database. To learn more, visit sigma.com/metlin.

Specifications

CAS 545-47-1
Molecular Formula C30H50O
Linear Formula C30H50O
MDL Number MFCD00017351
Quantity 25 mg
Synonym beta-Viscol; 20(29)-Lupen-3beta-ol; 3beta-Hydroxy-20(29)-lupene; Clerodol; Fagarasterol; Lupenol; Monogynol B
Molecular Weight (g/mol) 426.72 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.