Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lupeol, ≥90% (GC), MilliporeSigma™ Supelco™

Catalog No. 111014562 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-4562 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-4562 Supplier MilliporeSigma Supelco Supplier No. 1869210MG
Add to Cart
Add to Cart
Lupeol is a novel anti-cancer and anti-inflammatory dietary triterpene. They help in stabilizing phospholipid bilayers in plant cell membranes. In general, this group contains 28-29 carbons along with carbon-carbon double bonds, usually one in the sterol nucleus and at times a second in the alkyl side chain.
TRUSTED_SUSTAINABILITY

Specifications

CAS 545-47-1
Physical Form Neat
Linear Formula C30H50 O
MDL Number MFCD00017351
Synonym beta-Viscol; 20(29)-Lupen-3 beta-ol; 3 beta-Hydroxy-20(29)-lupene; Clerodol; Fagarasterol; Lupenol; Monogynol B
Type Analytical Standard
Quantity 10 mg
Formula Weight 426.72
Percent Purity ≥90% (GC)
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.