Learn More
Medchemexpress LLC Lupeol acetate | 1617-68-1 | MFCD00017362 | >98.0% | 468.75 | C32H52O2 | 5MG
SDP
Catalog No. 5000235023
Encompass_Preferred
Lupeol acetate is a triterpenoid derivative of lupeol used as an analytical standard and research reagent. It is reported to have anti-inflammatory, antibacterial, anti-trypanosomic, and anticancer activities and is used in bioactivity studies and analytical method development. The compound has CAS 1617-68-1, molecular formula C32H52O2, and molecular weight 468.75 g/mol.
  • Used as an analytical standard for chromatographic and reference purposes.
  • Reported biological activities include anti-inflammatory, antibacterial, anti-trypanosomic, and anticancer effects.
  • High chromatographic purity typically greater than 98% as supplied by quality vendors.
  • Chemical formula C32H52O2 and molecular weight 468.75 g/mol.
  • Suitable for research applications, bioactivity assays, and analytical method development.
Catalog No. 50-002-35023 Supplier Medchemexpress LLC Supplier No. HY1261145MG
Lupeol acetate is a triterpenoid derivative of lupeol used as an analytical standard and research reagent. It is reported to have anti-inflammatory, antibacterial, anti-trypanosomic, and anticancer activities and is used in bioactivity studies and analytical method development. The compound has CAS 1617-68-1, molecular formula C32H52O2, and molecular weight 468.75 g/mol.
  • Used as an analytical standard for chromatographic and reference purposes.
  • Reported biological activities include anti-inflammatory, antibacterial, anti-trypanosomic, and anticancer effects.
  • High chromatographic purity typically greater than 98% as supplied by quality vendors.
  • Chemical formula C32H52O2 and molecular weight 468.75 g/mol.
  • Suitable for research applications, bioactivity assays, and analytical method development.