Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Lupeol acetate | 1617-68-1 | MFCD00017362 | 98.0% | 468.75 g/mol | C32H52O2 | 10MG
SDP

Catalog No. 5000234962
Encompass_Preferred

Lupeol acetate is a triterpenoid acetate supplied as an analytical research standard for use in chemical and biological research. It serves as a reference material for analytical assays and research into anti-inflammatory, antibacterial, and anticancer activities. A certificate of analysis is provided for batch-specific storage and handling recommendations.

  • CAS number 1617-68-1
  • Chemical formula C32H52O2
  • Molecular weight 468.75 g/mol
  • Purity 98.0% (NMR)
  • Intended for use as an analytical standard and reference material
  • Certificate of analysis available for batch-specific storage and handling

Catalog No. 50-002-34962 Supplier Medchemexpress LLC Supplier No. HY12611410MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Lupeol acetate is a triterpenoid acetate supplied as an analytical research standard for use in chemical and biological research. It serves as a reference material for analytical assays and research into anti-inflammatory, antibacterial, and anticancer activities. A certificate of analysis is provided for batch-specific storage and handling recommendations.

  • CAS number 1617-68-1
  • Chemical formula C32H52O2
  • Molecular weight 468.75 g/mol
  • Purity 98.0% (NMR)
  • Intended for use as an analytical standard and reference material
  • Certificate of analysis available for batch-specific storage and handling

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.