Learn More
Medchemexpress LLC Lupeol (Clerodol; Monogynol B; Fagarasterol) | 545-47-1 | 426.72 | 25 MG
SDP
Catalog No. 5000389156
Encompass_Preferred
Lupeol is an active pentacyclic triterpenoid with anti-oxidant, anti-mutagenic, anti-tumor, and anti-inflammatory activities. It is also a potent androgen receptor (AR) inhibitor and is used for cancer research, particularly for androgen-dependent phenotype (ADPC) and castration-resistant phenotype (CRPC) prostate cancer.
  • Active pentacyclic triterpenoid
  • Anti-oxidant activity
  • Anti-mutagenic activity
  • Anti-tumor activity
  • Anti-inflammatory activity
  • Potent androgen receptor (AR) inhibitor
  • Can be used for cancer research, especially prostate cancer of ADPC and CRPC phenotypes
Catalog No. 50-003-89156 Supplier Medchemexpress LLC Supplier No. 25MGHYN0790
Lupeol is an active pentacyclic triterpenoid with anti-oxidant, anti-mutagenic, anti-tumor, and anti-inflammatory activities. It is also a potent androgen receptor (AR) inhibitor and is used for cancer research, particularly for androgen-dependent phenotype (ADPC) and castration-resistant phenotype (CRPC) prostate cancer.
  • Active pentacyclic triterpenoid
  • Anti-oxidant activity
  • Anti-mutagenic activity
  • Anti-tumor activity
  • Anti-inflammatory activity
  • Potent androgen receptor (AR) inhibitor
  • Can be used for cancer research, especially prostate cancer of ADPC and CRPC phenotypes