Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Luperox™ TBH70X, tert-Butyl hydroperoxide solution

Catalog No. 45813925ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mL
100 mL
1 L
4 L
18 L
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-813-925ML 25 mL
45-813-9100ML 100 mL
45-813-918L 18 L
45-813-91L 1 L
45-813-94L 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 45-813-925ML Supplier Sigma Aldrich Supplier No. 45813925ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Versatile reagent for oxidations.Product of Arkema Inc.

Specifications

CAS 75-91-2
Density 0.93 g/mL (at 25°C)
Boiling Point 96.2°C
Molecular Formula C4H10O2
Refractive Index n20/D 1.386
Linear Formula (CH3)3 COOH
Quantity 25 mL
MDL Number MFCD00002130
Synonym tert-Butyl hydroperoxide solution; TBHP; 1,1-Dimethylethyl hydroperoxide; 2-Hydroperoxy-2-methylpropane; TBHP
Molecular Weight (g/mol) 90.12
Percent Purity 12.25-14.20%active oxygen basis; 69.0-80.0% (NH2OH, titration)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.