Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Lupiwighteone | 104691-86-3 | 99.5% | 338.35 | C20H18O5 | 10 MG
SDP

Catalog No. 5000289623
Encompass_Preferred

Lupiwighteone is an isoflavone research compound used as a biochemical probe for studies of antioxidant, antimicrobial, and anticancer activity. It induces caspase-dependent and -independent apoptosis in cancer cells through inhibition of the PI3K/Akt/mTOR pathway. The material is supplied as a small solid suitable for preparation of DMSO stocks and in vivo formulation.

  • Isoflavone small molecule for in vitro and in vivo research.
  • Induces apoptosis via PI3K/Akt/mTOR pathway inhibition.
  • Solubility: DMSO 100 mg/mL (in vitro); ≥2.5 mg/mL in 10% DMSO/90% corn oil (in vivo).
  • Storage: store at 4°C, protect from light; in solvent -80°C (6 months) or -20°C (1 month).
  • Key identifiers: CAS 104691-86-3; formula C20H18O5; molecular weight 338.35.

Catalog No. 50-002-89623 Supplier Medchemexpress LLC Supplier No. HYN335410MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Lupiwighteone is an isoflavone research compound used as a biochemical probe for studies of antioxidant, antimicrobial, and anticancer activity. It induces caspase-dependent and -independent apoptosis in cancer cells through inhibition of the PI3K/Akt/mTOR pathway. The material is supplied as a small solid suitable for preparation of DMSO stocks and in vivo formulation.

  • Isoflavone small molecule for in vitro and in vivo research.
  • Induces apoptosis via PI3K/Akt/mTOR pathway inhibition.
  • Solubility: DMSO 100 mg/mL (in vitro); ≥2.5 mg/mL in 10% DMSO/90% corn oil (in vivo).
  • Storage: store at 4°C, protect from light; in solvent -80°C (6 months) or -20°C (1 month).
  • Key identifiers: CAS 104691-86-3; formula C20H18O5; molecular weight 338.35.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.