Learn More
Andwin Scientific LUQUINOX- 1 QT
SDP
Catalog No. NC0095803
Encompass
Liquinox yields reliable results with no interfering residues therefore ideal for cleaning oils, particulates and contaminants from anything from glassware, metals, plastic, ceramics, porcelain, rubber and fiberglass. It is completely soluble in hard and soft water, with a dilution ratio of 1:100. It has a pH level of 8.5. This is solid in quart packaging.
Catalog No. NC0095803 Supplier Andwin Scientific Supplier No. 12321
Liquinox yields reliable results with no interfering residues therefore ideal for cleaning oils, particulates and contaminants from anything from glassware, metals, plastic, ceramics, porcelain, rubber and fiberglass. It is completely soluble in hard and soft water, with a dilution ratio of 1:100. It has a pH level of 8.5. This is solid in quart packaging.