Learn More
Apexbio Technology LLC Lurasidone HCl,10mg CAS# 367514-88-3
SDP
Supplier:  Apexbio Technology LLC B147710
Encompass_Preferred
Lurasidone Hcl (SM 13496 Hcl) is an inhibitor of Dopamine D2, 5-HT2A, 5-HT7, 5-HT1A and noradrenaline α2C with IC50 of 1.68 nM, 2.03 nM, 0.495 nM, 6.75 nM and 10.8 nM, respectively, which is an atypical antipsychotic. Other sizes are also available. Please inqury us for quote.
Catalog No. 50-224-0366