Learn More
TARGETMOL CHEMICALS INC LURASIDONE HYDROCHLORIDE 100MG
SDP
Catalog No. 502581105
Encompass_Preferred
Also available in 5 mg 10 mg 25 mg 50 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Lurasidone hydrochloride (Lurasidone HCl) is a thiazole derivative and atypical antipsychotic agent that functions as a dopamine D2 receptor antagonist; serotonin 5-HT2 receptor antagonist serotonin 5-HT7 receptor antagonist an antagonist of the adrenergic (alpha)2A and (alpha)2C receptors as well as a partial serotonin 5-HT1A receptor agonist. It is used in the treatment of schizophrenia and bipolar disorder. purity: 99%
Catalog No. 50-258-1105 Supplier TARGETMOL CHEMICALS INC Supplier No. T1735100MG
Also available in 5 mg 10 mg 25 mg 50 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Lurasidone hydrochloride (Lurasidone HCl) is a thiazole derivative and atypical antipsychotic agent that functions as a dopamine D2 receptor antagonist; serotonin 5-HT2 receptor antagonist serotonin 5-HT7 receptor antagonist an antagonist of the adrenergic (alpha)2A and (alpha)2C receptors as well as a partial serotonin 5-HT1A receptor agonist. It is used in the treatment of schizophrenia and bipolar disorder. purity: 99%