Learn More
Medchemexpress LLC Lurasidone hydrochloride | 367514-88-3 | 99.86% | C28H37ClN4O2S | 100 MG
SDP
Catalog No. 5000432756
Encompass_Preferred
Lurasidone hydrochloride is a chemical reagent from MedChemExpress that functions as an antagonist for both dopamine D2 and 5-HT7 receptors. It also acts as a partial agonist of the 5-HT1A receptor.
  • Antagonist of dopamine D2 and 5-HT7 receptors
  • Partial agonist of 5-HT1A receptor
  • Suitable for neurological disease research
  • Stable under recommended storage conditions
Catalog No. 50-004-32756 Supplier Medchemexpress LLC Supplier No. HYB0032100MG
Add to Cart
Add to Cart
Lurasidone hydrochloride is a chemical reagent from MedChemExpress that functions as an antagonist for both dopamine D2 and 5-HT7 receptors. It also acts as a partial agonist of the 5-HT1A receptor.
  • Antagonist of dopamine D2 and 5-HT7 receptors
  • Partial agonist of 5-HT1A receptor
  • Suitable for neurological disease research
  • Stable under recommended storage conditions