Learn More
Selleck Chemical LLC Lurbinectedin1mg
SDP
Supplier:  Selleck Chemical LLC S96031MG
Encompass_Preferred
Lurbinectedin (PM01183, PM-1183, LY-01017, Tryptamicidin, Zepsyre, ZEPZELCA), aDNA minor groove covalent binder with potent in vitro and in vivo anti-tumour activity. Lurbinectedin inhibits RMG1 and RMG2 cell growth with IC50 of 1.25 nM and 1.16 nM, respectively. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3474