Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Research Products International Corp Luria Agar 500 Milliliter Buffered Capsules [Miller's LB Agar], 1 Kilogram
SDP

Catalog No. 50550415
Encompass_Preferred

RPI media is now available in convenient premeasured and pre-mixed capsules. Prebuffered media eliminates pH adjustments. Capsules contain all necessary ingredients for maintenance and propagation of E. coli.

Simply add 2 x 500 mL capsules to one liter of water, sterilize by autoclaving for 15 minutes at 121°C. Supplied as 40 gm capsule.

Final Solution Contains:

Tryptone: 10 g/l
Yeast Extract: 5 g/l
Sodium Choride: 10 g/l
Agar: 15 g/l
Tris/Tris HCL: 1.5 g/l

Catalog No. 50-550-415 Supplier Research Products International Corp Supplier No. L240251000.0
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

RPI media is now available in convenient premeasured and pre-mixed capsules. Prebuffered media eliminates pH adjustments. Capsules contain all necessary ingredients for maintenance and propagation of E. coli.

Simply add 2 x 500 mL capsules to one liter of water, sterilize by autoclaving for 15 minutes at 121°C. Supplied as 40 gm capsule.

Final Solution Contains:

Tryptone: 10 g/l
Yeast Extract: 5 g/l
Sodium Choride: 10 g/l
Agar: 15 g/l
Tris/Tris HCL: 1.5 g/l

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.