Learn More
Bioworld Luria Agar Base, Miller, 1 kg
Catalog No. 50254431
Encompass_Preferred
Luria Agar BaseFormulated for the propagation and maintenance of E. coli for molecular biologyContains one-tenth the sodium chloride content as LB Agar, Lennox and one-twentieth the sodium chloride content as LB Agar, Miller formulationspH: 7.0Prepared Appearance: light amber agarComposition:Bacto Tryptone Bacto Yeast Extract Sodium Chloride Bacto Agar
Catalog No. 50-254-431 Supplier Bioworld Supplier No. 306210122
Luria Agar BaseFormulated for the propagation and maintenance of E. coli for molecular biologyContains one-tenth the sodium chloride content as LB Agar, Lennox and one-twentieth the sodium chloride content as LB Agar, Miller formulationspH: 7.0Prepared Appearance: light amber agarComposition:Bacto Tryptone Bacto Yeast Extract Sodium Chloride Bacto Agar