Learn More
Bioworld Luria Agar Base, Miller 500g
Catalog No. 50254430
Encompass_Preferred
Formulated for the propagation and maintenance of E. coli for molecular biology. Contains one-tenth the sodium chloride content as LB Agar, Lennox and one-twentieth the sodium chloride content as LB Agar, Miller formulations. pH: 7.0, Prepared Appearance: light amber agar. Composition: Bacto Tryptone, Bacto Yeast Extract, Sodium Chloride, Bacto Agar, Solubility : Soluble in boiling water, Appearance Form : Powder, Appearance Color : Light tan. 500g
Catalog No. 50-254-430 Supplier Bioworld Supplier No. 306210121
Formulated for the propagation and maintenance of E. coli for molecular biology. Contains one-tenth the sodium chloride content as LB Agar, Lennox and one-twentieth the sodium chloride content as LB Agar, Miller formulations. pH: 7.0, Prepared Appearance: light amber agar. Composition: Bacto Tryptone, Bacto Yeast Extract, Sodium Chloride, Bacto Agar, Solubility : Soluble in boiling water, Appearance Form : Powder, Appearance Color : Light tan. 500g