Learn More
Alkali Scientific Luria Agar [Miller's LB] 1 Gram Buffered Capsules, 500 G
SDP
Supplier:  Alkali Scientific URI002CP
Encompass_Preferred
Luria Agar [Miller's LB] 1 Gram Buffered Capsules, 500G is a microbial culture media used in a variety of laboratory applications, including bacterial cultivation and microbial research. The formulation contains a balanced nutrient mix that supports the growth of a wide range of microorganisms. These capsules are designed to be buffered, ensuring a stable pH environment that is crucial for consistent and reliable results. The 500g bottle provides a convenient volume for laboratories requiring moderate amounts of prepared media. The media is commonly used in microbiology labs for culturing bacteria, yeast, and fungi. It is available as a buffered capsule that can be easily dissolved in water.
Catalog No. 50-006-61695