Learn More
Alkali Scientific Luria Agar [Miller's LB] Powder, 2 Kg
SDP
Supplier:  Alkali Scientific RMI003CP
Encompass
Luria Agar (Miller's LB) Powder is a widely used growth medium formulated for cultivating bacteria, such as Escherichia coli, in laboratory settings. Available in a powdered form, this medium is easily prepared by dissolving in water. It is ideal for research and industrial applications, supporting bacterial growth by providing essential nutrients such as nitrogen, carbon, and vitamins. This 2kg size is suitable for medium-sized laboratories requiring frequent media preparation. Luria Agar is used extensively in microbiology and molecular biology research, particularly in studies involving plasmid transformation, bacterial culture, and colony formation. This medium supports both general and selective bacterial growth.
Catalog No. 50-006-61676