Learn More
Alkali Scientific Luria Agar [Miller's LB] Powder, 5 Kg
SDP
Catalog No. 5000661746
Encompass_Preferred
Luria Agar (Miller's LB) Powder is a comprehensive bacterial growth medium used in microbiology and molecular biology research. The powdered form is easy to prepare by simply dissolving it in water, making it ideal for growing microorganisms like Escherichia coli. This 5kg size is suitable for medium to large-scale laboratories requiring frequent media preparation. Luria Agar supports bacterial growth by providing essential nutrients such as nitrogen, carbon, salts, and vitamins. It is used in a wide range of applications, including plasmid cloning, bacterial transformation, and microbial studies. This product is a versatile solution for routine and specialized bacterial growth protocols.
Catalog No. 50-006-61746 Supplier Alkali Scientific Supplier No. RMI006CP
Luria Agar (Miller's LB) Powder is a comprehensive bacterial growth medium used in microbiology and molecular biology research. The powdered form is easy to prepare by simply dissolving it in water, making it ideal for growing microorganisms like Escherichia coli. This 5kg size is suitable for medium to large-scale laboratories requiring frequent media preparation. Luria Agar supports bacterial growth by providing essential nutrients such as nitrogen, carbon, salts, and vitamins. It is used in a wide range of applications, including plasmid cloning, bacterial transformation, and microbial studies. This product is a versatile solution for routine and specialized bacterial growth protocols.