Learn More
Bioworld Luria-Bertani (LB) Agar, Lennox, 1 kg
Catalog No. 50254312
Encompass_Preferred
Luria-Bertani (LB) Agar, LennoxFor propagation and maintenance of Escherichia coliLow salt formulationpH: 7.0Working Concentration: 35 g/LDehydrated Appearance: light beige, homogeneous powderPrepared Appearance: medium amber agarSoluble in boiling waterContains:TryptoneYeast ExtractSodium ChlorideAgarThis product is sold for laboratory research use only.
Catalog No. 50-254-312 Supplier Bioworld Supplier No. 306200352
Luria-Bertani (LB) Agar, LennoxFor propagation and maintenance of Escherichia coliLow salt formulationpH: 7.0Working Concentration: 35 g/LDehydrated Appearance: light beige, homogeneous powderPrepared Appearance: medium amber agarSoluble in boiling waterContains:TryptoneYeast ExtractSodium ChlorideAgarThis product is sold for laboratory research use only.