Learn More
Bioworld Luria-Bertani (LB) Agar, Miller, 20-Gram Capsules, 25 Capsule(s)
Catalog No. 50254416
Encompass_Preferred
Luria-Bertani (LB) Agar, MillerFor propagation and maintenance of Escherichia coliConvenient premeasured and pre-mixed capsulesContains twenty times the sodium chloride level of Luria Agar Base, Miller and twice the level found in LB Agar, LennoxpH: 7.0Working Concentration: 40 g/L (2 capsules)Prepared Appearance: light amber agarSoluble in boiling waterContains:TryptoneYeast ExtractSodium ChlorideAgarThis product is sold for laboratory research use only.
Catalog No. 50-254-416 Supplier Bioworld Supplier No. 306201131
Luria-Bertani (LB) Agar, MillerFor propagation and maintenance of Escherichia coliConvenient premeasured and pre-mixed capsulesContains twenty times the sodium chloride level of Luria Agar Base, Miller and twice the level found in LB Agar, LennoxpH: 7.0Working Concentration: 40 g/L (2 capsules)Prepared Appearance: light amber agarSoluble in boiling waterContains:TryptoneYeast ExtractSodium ChlorideAgarThis product is sold for laboratory research use only.