Learn More
Bioworld Luria-Bertani (LB) Agar, Miller, 2 kg
Catalog No. 50254323
Encompass_Preferred
Luria-Bertani (LB) Agar, MillerFor propagation and maintenance of Escherichia coliContains twenty times the sodium chloride level of Luria Agar Base, Miller and twice the level found in LB Agar, LennoxpH: 7.0Working Concentration: 37 g/LDehydrated Appearance: light tan, homogeneous powderPrepared Appearance: light amber agarSoluble in boiling waterContains:TryptoneYeast ExtractSodium ChlorideAgar Usually ships within 24 hours. This product is sold for laboratory research use only.
Catalog No. 50-254-323 Supplier Bioworld Supplier No. 306200424
Luria-Bertani (LB) Agar, MillerFor propagation and maintenance of Escherichia coliContains twenty times the sodium chloride level of Luria Agar Base, Miller and twice the level found in LB Agar, LennoxpH: 7.0Working Concentration: 37 g/LDehydrated Appearance: light tan, homogeneous powderPrepared Appearance: light amber agarSoluble in boiling waterContains:TryptoneYeast ExtractSodium ChlorideAgar Usually ships within 24 hours. This product is sold for laboratory research use only.