Learn More
Bioworld Luria-Bertani (LB) Broth, Lennox, 20-Gram Capsules, 1 kg
Catalog No. 50254315
Encompass_Preferred
Luria-Bertani (LB) Broth, LennoxFor propagation and maintenance of Escherichia coliConvenient premeasured and pre-mixed capsulesContains ten times the sodium chloride level of Luria Broth Base, Miller and one half of that found in Luria-Bertani (LB) Broth, MillerpH: 7.0Working Concentration: 20 g/L (1 capsule)Prepared Appearance: light amber brothSoluble in waterContains:TryptoneYeastSodium ChorideThis product is sold for laboratory research use only.
Catalog No. 50-254-315 Supplier Bioworld Supplier No. 306200362
Luria-Bertani (LB) Broth, LennoxFor propagation and maintenance of Escherichia coliConvenient premeasured and pre-mixed capsulesContains ten times the sodium chloride level of Luria Broth Base, Miller and one half of that found in Luria-Bertani (LB) Broth, MillerpH: 7.0Working Concentration: 20 g/L (1 capsule)Prepared Appearance: light amber brothSoluble in waterContains:TryptoneYeastSodium ChorideThis product is sold for laboratory research use only.